Potassium hydroxide 1 N in aqueous solution, BAKER ANALYZED® ACS volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = NI

BAKR5602-03 5602-02
CAJT5602-3CS 1144.03 CAD
CAJT5602-3 JT5602-2
Potassium hydroxide 1 N in aqueous solution, BAKER ANALYZED® ACS volumetric solution, J.T.Baker®
Potassium hydroxide

Standardized at 25 °C against chemicals whose certification is traceable to NIST.

Formula: KOH
Density: 1.05 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.995 - 1.005
Chloride (Cl) ≤10 ppm
Nitrogen Compounds (as N) ≤1 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Trace Impurities - Iron (Fe) ≤0.5 ppm
Appearance Passes Test
Standardization at 25 °C traceable to NIST Standard Reference Material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR