FlexTech™ Stretch Gloves with Foam Nitrile Palm Coating, Wells Lamont
Supplier: Wells Lamont
Y9256XS Y9256L Y9256M Y9256S
89030-808PK 284.59 CAD
89030-808 80092-386 80092-388 80092-390 80092-392
FlexTech™ Stretch Gloves with Foam Nitrile Palm Coating, Wells Lamont
Gloves
These gloves are ideal for general assembly, automotive industry, sharp or pointy small parts, and component and glass handling.
  • Ideal for handling sharp objects
  • Lightweight
  • Protection with comfort
  • Seamless design for a close fit
  • Easy to wash
Stretchy Kevlar®/Lycra® blend provides high level of dexterity and comfort, and foam nitrile palm provides excellent grip.
The easy-to-wash gloves provide protection without sacrificing dexterity or comfort. The color-coded wrist makes size identification fast.