Petroleum spirit, 35…60 °C, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

BAKR9268-05 CAJT9268-7 BAKR9268-22 9268-03 BAKR9268-05 BAKR9268-09
CAJT9268-5CS 1201.07 CAD
CAJT9268-5 CAJT9268-7 CAJT9268-22 CAJT9268-3 JT9268-3 JT9268-05 CAJT9268-9
Petroleum spirit, 35…60 °C, BAKER ANALYZED® ACS, J.T.Baker®
Petroleum spirit
Boiling Pt: 35 - 60 °C (1013 hPa)
Density: ~0.68 kg (20 °C)
Flash Pt: −18 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤0.001 %
ACS - Acidity Passes Test
ACS - Boiling Range(Lower) 35 - 60 °C
ACS - Boiling Range(Upper) 35 - 60 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR