1-Propanol ≥99.0% (by GC, corrected for water content), BAKER ANALYZED®, J.T.Baker®

Supplier: VWR International
Danger

Synonyms: Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane

Management of Change (MOC) category = N

BAKR9086-07 BAKR9086-01 BAKR9086-03 BAKR9086-33 9086-05
CAJT9086-7EA 1527.04 CAD
CAJT9086-7 CAJT9086-1 CAJT9086-3 CAJT9086-33 CAJT9086-5 JT9086-5
1-Propanol ≥99.0% (by GC, corrected for water content), BAKER ANALYZED®, J.T.Baker®
1-Propanol

BAKER ANALYZED™ Reagent—A grade of high-quality chemicals for laboratory and specialized industrial use.

Formula: H₃CCH₂CH₂OH
MW: 60.1 g/mol
Boiling Pt: 97
Melting Pt: –127 °C
Density: 0.8053 g/cm³ (20 °C)
Flash Pt: 15 (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002941
CAS Number: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3 II
Merck Index: 13,07934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (CH₃CH₂CH₂OH) (by GC, corrected for water) ≥99.0 %
Residue after Evaporation ≤0.0100 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR