2-Propanol 99,5, BAKER ANALYZED® A.C.S. Reagent ACS, J.T.Baker®

Supplier: VWR International
Danger

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

M.O.C. Category N

CAJT9084-7 BAKR9084-R BAKR9084-22 BAKR9084-33 BAKR9084-05 9084-06 9084-03
CAJT9084-7EA 1174.59 CAD
CAJT9084-7 CAJT9084-R CAJT9084-1 CAJT9084-22 CAJT9084-3 CAJT9084-33 CAJT9084-5 CAJT9084-6 JT9084-6 JT9084-3
2-Propanol 99,5, BAKER ANALYZED® A.C.S. Reagent ACS, J.T.Baker®
2-Propanol

The quality of many BAKER ANALYZED™ Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These solvents are essential for any chemical stockroom.

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (CH₃CHOHCH₃) ≥ 99.5 %
Carbonyl Compounds - Acetone ≤ 0.002 %
Carbonyl Compounds - Propionaldehyde ≤ 0.002 %
Color (APHA) ≤ 10
Residue after Evaporation ≤ 0.001 %
Solubility in H₂O Passes Test
Titrable Acid or Base (meq/g) ≤ 0.0001
Water (by KF, coulometric) ≤ 0.2 %
Trace Impurities - Copper (Cu) ≤ 0.10 ppm
ACS - Heavy Metals (as Pb) ≤ 1 ppm
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Trace Impurities - Nickel (Ni) ≤ 0.1 ppm
Date Received: __ __ /__ __ /__ __
Date Opened: __ __ /__ __ /__ __
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR