2-Propanol ≥99.5%, PHOTREX® for UV spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

Management of Change (MOC) category = N

BAKR9083-01 9083-03
CAJT9083-1CS 1911.48 CAD
CAJT9083-1 CAJT9083-3 JT9083-3
2-Propanol ≥99.5%, PHOTREX® for UV spectrophotometry, J.T.Baker®
2-Propanol

Pure, trusted chromatography solvents providing high selectivity, reproducibility and accuracy of results that come in a variety of grades such as HPLC, LCMS, ULTRA LCMS and PHOTREX.

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (CH₃CHOHCH₃) ≥99.5 %
Carbonyl Compounds - Acetone ≤0.002 %
Carbonyl Compounds - Propionaldehyde ≤0.002 %
Color (APHA) ≤10
Residue after Evaporation ≤0.0005 %
Solubility in H₂O Passes Test
ACS - Titrable Acid or Base (µeq/g) ≤0.1
Water (by KF, coulometric) ≤0.05 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-330 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 300 nm ≤0.02
Ultraviolet Absorbance (1.00-cm cell vs. water) - 275 nm ≤0.03
Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm ≤0.04
Ultraviolet Absorbance (1.00-cm cell vs. water) - 245 nm ≤0.08
Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm ≤0.20
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm ≤0.40
Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm ≤1.00
Product Information (not specifications):
Boiling Point (typical)................. 82.3° C.
Density (g/mL) at 25° C (typical)........ 0.782
Windows of Infrared Transmittance (0.1-mm path, 50-100% T), µm:
2.5 - 2.8; 4.1 - 5.6; 5.8 - 6.4
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR