Heat Sealer, Ampac®
Supplier: Ampac Flexibles
118-1 101-6
11214-150EA 2773.28 CAD
11214-150 11214-107
Heat Sealer, Ampac®
Heat Sealers Bag Sealers
Ampac's 11" and 18" heat sealers are excellent for use with all Ampac pouches
They provide airtight seals and are thermostat controlled to a maintain a temperature of approximately 167.8°C (325°F). Mounts on wall or under cabinet as well as bench top. Requires 3 minutes to preheat, and 5 seconds to seal.
Caution: Not for use with plastic/polyethylene bags. Intended for multi-ply barrier laminations. For best results, use only Ampac pouches.
Order Now
SPECIFICATIONS
Electrical 115V
Environmentally Preferable