2-Propanol ≥99% USP for biochemistry, for cleaning and processing, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = R

BAKR9080-07 BAKR9080-R BAKR9080-01 9080-03
CAJT9080-7EA 1413.77 CAD
CAJT9080-7 CAJT9080-R CAJT9080-1 CAJT9080-3 JT9080-3
2-Propanol ≥99% USP for biochemistry, for cleaning and processing, J.T.Baker®

Achieve unsurpassed consistency, accurate results and industry-leading performance with solvents from Avantor™ Performance Materials. J.T.Baker® and Macron Fine Chemicals™ brands of solvents are available in sizes ranging from 500 mL to bulk packages containing hundreds of gallons. Our bulk solvent delivery capabilities are ideal for both pharmaceutical and biopharmaceutical production.

Formula: (CH₃)₂CHOH
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets U.S.P Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Specific Gravity at 25°/25°C 0.783 - 0.787
USP - Refractive Index, n²ºD 1.376 - 1.378
USP - Acidity Passes Test
USP - Nonvolatile Residue ≤0.005 %
USP - Assay ≥99.0 %
USP - Water (H₂O) ≤0.5 %
USP - Volatile Impurities - Ethyl ether ≤0.1 %
USP - Volatile Impurities - Acetone ≤0.1 %
USP - Volatile Impurities - Diisopropyl ether ≤0.1 %
USP - Volatile Impurities - 1-Propanol ≤0.1 %
USP - Volatile Impurities - 2-Butanol ≤0.1 %
USP - Volatile Impurities - Total ≤1.0 %
USP - Benzene ≤2 ppm
Typical Property: Appearance (clear, colorless liquid)
Residual solvents – Only the Class 2 solvent cyclohexane and Class 3
solvents are likely to be present. Cyclohexane is controlled below the
USP 467 Option 1 limit. All Class 3 solvents are less than 0.5%.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR