TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.0% Made from Tromethamine USP, Biotech Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Tris(hydroxymethyl)methylamine hydrochloride, 2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride, Trizma® hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, TRIS HCl, Tromethane hydrochloride, Tris-HCl, Tris(hydroxymethyl)methylammonium chloride, Trometamol hydrochloride, THAM HCl

Management of Change (MOC) category = R

BAKR4106-01 BAKR4106-05 BAKR4106-09 BAKR4106-F9 4106-F7 BAKR4106-F7
CAJT4106-1EA 808.17 CAD
CAJT4106-1 CAJT4106-5 CAJT4106-7 CAJT4106-9 CAJT4106-F9 JT4106-F7 CAJT4106-F7
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.0% Made from Tromethamine USP, Biotech Reagent, J.T.Baker®
TRIS HCl (Tris-(hydroxymethyl) aminomethane hydrochloride)

GMP manufactured product. The infrared spectrum conforms to the standard.

Formula: NH₂C(CH₂OH)₃·HCl
MW: 157.6 g/mol
Melting Pt: 147…153 °C
Density: 1.05 g/ml (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012590
CAS Number: 1185-53-1
EINECS: 214-684-5
Merck Index: 12,09902
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cbdc9446fdb9bcfe3f1d992ff20a16726f98113&vid=3d668801395b81fa595401a752279006da8fd3cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Assay (NH₂C(CH₂OH)₃·HCl) ≥99.0 %
Appearance (colorless crystals to white crystalline powder) Passes Test
Absorbance of a 1M Solution - 260 nm ≤0.06
Absorbance of a 1M Solution - 280 nm ≤0.06
Absorbance of a 1M Solution - 400 nm ≤0.01
Identification (by IR) Passes Test
Loss on Drying at 105°C ≤0.5 %
Endotoxin Concentration (EU/g) ≤2.5
Melting Point 147 - 153 °C
pH of 0.5M Solution at 25°C 3.5 - 5.0
Trace Impurities - Arsenic (As) ≤10 ppm
Trace Impurities - Calcium (Ca) ≤10.0 ppm
Trace Impurities - Copper (Cu) ≤10 ppm
Trace Impurities - Iron (Fe) ≤10 ppm
Trace Impurities - Lead (Pb) ≤10 ppm
Trace Impurities - Magnesium (Mg) ≤10 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Manufactured exclusively for Avantor Performance Materials, LLC., by BioSpectra, Inc., Stroudsburg, PA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR