European Style Five-Neck Flasks, Jacketed, Ace Glass
Supplier: Ace Glass
6959-44 6959-48 6959-51 6959-55
89083-842EA 1414.27 CAD
89083-842 89083-844 89083-846 89083-848
European Style Five-Neck Flasks, Jacketed, Ace Glass
Flasks Round Bottom Flasks
European style flasks have tapered walls with a shallow bottom for more efficient magnetic or mechanical half-moon shaped paddle stirring
The full jacket has two barbed side tubes for hoses. These five-neck flasks have three [ST] outer joints and two #7 Ace-Thred necks. The outer joints are tooled with reinforcement rings for added strength and stability. Two of the side necks are in-line with the center neck. The remaining side necks are placed in front of the in-line necks and are internally threaded to accommodate thermometers or bleed tubes up to 7mmO.D. Supplied with two #7 nylon bushings and FETFE O-rings.