TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99% (dried basis), Ultrapure Bioreagent for liquid chromatography, for LC-MS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Synonyms:
Tris(hydroxymethyl)methylamine hydrochloride, 2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride, Trizma® hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, TRIS HCl, Tromethane hydrochloride, Tris-HCl, Tris(hydroxymethyl)methylammonium chloride, Trometamol hydrochloride, THAM HCl
Management of Change (MOC) category = N
BAKR4103-01
BAKR4103-02
BAKR4103-06
BAKR4103-07
BAKR4103-08
BAKR4103-F7
BAKR4103-04
CAJT4103-1EA
620.69
CAD
CAJT4103-1
CAJT4103-2
CAJT4103-6
CAJT4103-7
CAJT4103-8
CAJT4103-F7
CAJT4103-4
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99% (dried basis), Ultrapure Bioreagent for liquid chromatography, for LC-MS, J.T.Baker®
TRIS HCl (Tris-(hydroxymethyl) aminomethane hydrochloride)
For liquid chromatography and molecular biology applications.
- Lot analysis on label
Formula:
NH₂C(CH₂OH)₃·HCl Melting Pt: 150…152 °C Density: 1.05 g/ml (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00012590 CAS Number: 1185-53-1 EINECS: 214-684-5 Merck Index: 12,09902 |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (NH₂C(CH₂OH)₃·HCl) (dried basis) | ≥99.0 % |
Appearance | Passes Test |
Absorbance of a 1M Solution - 260 nm | ≤0.06 |
Absorbance of a 1M Solution - 280 nm | ≤0.06 |
Absorbance of a 1M Solution - 400 nm | ≤0.01 |
Water (H₂O)(by Karl Fischer titrn) | ≤0.5 % |
RNase Activity (None Detected) | Passes Test |
DNase Activity (None Detected) | Passes Test |
Protease Activity (None Detected) | Passes Test |
Trace Impurities - Calcium (Ca) | ≤1 ppm |
Trace Impurities - Copper (Cu) | ≤1 ppm |
Trace Impurities - Iron (Fe) | ≤1 ppm |
Trace Impurities - Lead (Pb) | ≤1 ppm |
Trace Impurities - Magnesium (Mg) | ≤1.000 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...