Talc, powder, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = R

BAKR4100-05 BAKR4100-R 4100-01
CAJT4100-5EA 340.03 CAD
CAJT4100-5 CAJT4100-R CAJT4100-1 JT4100-1
Talc, powder, Multi-Compendial, J.T.Baker®
Talc

White to off-white fine powder.

Formula: Mg₃O₁₁Si₄·1H₂O
Melting Pt: 1200 °C
Density: 2.58…2.83 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00792903
CAS Number: 14807-96-6
EINECS: 238-877-9
Merck Index: 13,09127

Order Now

Specification Test Results

GMP Manufactured Product
Meets U.S.P Requirements
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C Passes Test
Microbial Limit - Total Aerobic Bacterial Count ≤100 cfu/g
Microbial Limit - Total Molds and Yeasts ≤50 cfu/g
USP - Acidity and Alkalinity Passes Test
USP - Loss on Ignition ≤7.0 %
USP - Water-Soluble Substances ≤0.1 %
USP - Iron (Fe) ≤0.25 %
USP - Lead (Pb) ≤0.001 %
USP - Calcium (Ca) ≤0.9 %
USP - Aluminum (Al) ≤2.0 %
USP - Absence of Asbestos (Method A) Passes Test
USP - Magnesium (as MgO) 17.0 - 19.5 %
EP/BP - Free from Asbestos Passes Test
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Water-Soluble Substances ≤0.2 %
EP/BP - Aluminum (Al) ≤2.0 %
EP/BP - Calcium (Ca) ≤0.90 %
EP/BP - Iron (Fe) ≤0.25 %
EP/BP - Lead (Pb) ≤10.0 ppm
EP/BP - Magnesium (Mg) 17.0 - 19.5 %
EP/BP - Loss on Ignition ≤7.0 %
EP/BP - Total Aerobic Microbial Count, cfu/g ≤1000
EP/BP - Total Yeast and Mold Count, cfu/g ≤100
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Suitable for Oral Administration

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR