European Style Three-Neck Flasks, Angled Necks, Jacketed, Ace Glass
Supplier: Ace Glass
6959-30 6959-33 6959-03 6959-05 6959-36 6959-38 6959-11 6959-13 6959-22 6959-24 6959-26
89083-820EA 1639.54 CAD
89083-820 89083-822 89083-824 89083-826 89083-828 89083-830 89083-832 89083-834 89083-836 89083-838 89083-840
European Style Three-Neck Flasks, Angled Necks, Jacketed, Ace Glass
Flasks Round Bottom Flasks
European style flasks have tapered walls with a shallow bottom for more efficient magnetic or mechanical half-moon shaped paddle stirring
The vertical center neck and angled side necks have [ST] outer joints that are tooled with reinforcement rings for added strength and stability. The full jacket has two barbed side tubes for hoses.