Trichloroacetic acid (TCA) 99,0 Acidimetry, crystals, BAKER ANANYZED™ ACS

Supplier: VWR International
Danger

Synonyms: TCA

M.O.C. Category N

CAJT0414-4 BAKR0414-01
CAJT0414-4EA 267.89 CAD
CAJT0414-4 CAJT0414-1 CAJT0414-5
Trichloroacetic acid (TCA) 99,0 Acidimetry, crystals, BAKER ANANYZED™ ACS
Trichloroacetic acid (TCA)

Crystals. Lot analysis on label.


  • Lot analysis on label

Formula: Cl₃CCOOH
MW: 163.39 g/mol
Boiling Pt: 196
Melting Pt: 54…56 °C
Density: 1.63 g/cm³ (20 °C)
Flash Pt: 113 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
Merck Index: 13,09700
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
IMPORTANT: Store or transport at or below 30°C (86°F).
Assay (Cl₃CCOOH) (by acidimetry) ≥99.0 %
Clarity of Solution Passes Test
Insoluble Matter ≤0.01 %
Residue after Ignition ≤0.03 %
Chloride (Cl) ≤0.002 %
Nitrate (NO₃) ≤0.002 %
ACS - Phosphate (PO₄) ≤5 ppm
Sulfate (SO₄) ≤0.02 %
Heavy Metals (as Pb) ≤0.002 %
Trace Impurities - Iron (Fe) ≤0.001 %
Substances Darkened by H₂SO₄ Passes Test
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR