Tetrahydrofuran 99,8 no preservatives_contains ACS, meetSpec_USP HPLC, Spectrophotometry
Supplier: HONEYWELL BURDICK & JACKSON CA
|
Danger
|
Synonyms:
Butylene oxide, THF, Diethylene oxide
AH340-4PC
AH340-4
CABJAH340-4PCCS
1953.2
CAD
CABJAH340-4PC
CABJAH340-4
Tetrahydrofuran 99,8 no preservatives_contains ACS, meetSpec_USP HPLC, Spectrophotometry
Tetrahydrofuran
Meets USP Reagent Specifications
- ACS certified
- Lot-to-lot consistency
- HPLC grade controlled for UV absorbance, GC assay, water content and non-volatile residue
Formula:
C₄H₈O MW: 72.11 g/mol Boiling Pt: 66 Melting Pt: -108.5 Density: 0.8892 g/cm³ (20 °C) Flash Pt: –14 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005356 CAS Number: 109-99-9 EINECS: 203-726-8 UN: 2056 ADR: 3 II REACH: 05-2114615188-48 Merck Index: 12,09351 |
|
Specification Test Results
Color (APHA) | 20 max. |
GC Analysis | 99.8 % min. |
Peroxides (as H₂O₂) at time of packaging | 2 mg/L max. |
Refractive Index (20ºC) | 1.4065 - 1.4075 |
Residue | 3 mg/L max. |
UV Absorbance @ 250 nm | 0.18 AU max. |
UV Absorbance @ 300 nm | 0.02 AU max. |
UV Cutoff | 212 nm max. |
Water by Karl Fischer Titration | 0.03 % max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...