Sodium Dimethyl 5-sulfoisophthalate ≥98.0%

Supplier: TCI America

Synonyms: Dimethyl sodium 5-sulfoisophthalate, Dimethyl 5-sulfoisophthalate sodium salt, 5-Sulphoisophthalic acid dimethyl ester sodium salt, Sodium 3,5-bis(methoxycarbonyl)benzenesulphonate, Sodium 3,5-bis(methoxycarbonyl)benzenesulfonate, Dimethyl 5-sulphoisophthalate sodium salt, Dimethyl sodium 5-sulphoisophthalate, 5-Sulfoisophthalic acid dimethyl ester sodium salt

S0157-500G S0157-25G
TCS0157-500GEA 232.26 CAD
TCS0157-500G TCS0157-025G
Sodium Dimethyl 5-sulfoisophthalate ≥98.0%
Sodium Dimethyl 5-sulfoisophthalate

CAS Number: 3965-55-7
MDL Number: MFCD00007493
Molecular Formula: C10H10O7S
Molecular Weight: 296.22
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Flash Point (°C): 200


Product Categories: Materials Chemistry, Anionic Surfactants
Other Product Names / Synonyms: Dimethyl Sodium 5-Sulfoisophthalate, 5-Sulfoisophthalic Acid Dimethyl Ester Sodium Salt, Dimethyl 5-Sulfoisophthalate Sodium Salt

Formula: C₁₀H₁₀O₇S
MW: 296.22 g/mol
Flash Pt: 200 °C
MDL Number: MFCD00007493
CAS Number: 3965-55-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1b386637b5c5d007ceacadc4aa9248c8929f274&vid=a1b386637b5c5d007ceacadc4aa9248c8929f274&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR