di-Sodium succinate hexahydrate ≥98.0%

Supplier: TCI America

Synonyms: Sodium succinate hexahydrate, Succinic acid disodium hexahydrate, 1,4-Butanedioic acid disodium salt hexahydrate, Sodium Succinate, Hexahydrate, Sodium succinate dibasic hexahydrate, Disodium succinate hexahydrate, Succinic acid disodium salt hexahydrate

S0105-25G S0105-500G
TCS0105-025GEA 62.66 CAD
TCS0105-025G TCS0105-500G
di-Sodium succinate hexahydrate ≥98.0%
di-Sodium succinate hexahydrate

CAS Number: 6106-21-4
MDL Number: MFCD00002790
Molecular Formula: C4H6O4
Molecular Weight: 162.05
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Other Product Names / Synonyms: Succinic Acid Disodium Salt

Formula: NaOOCCH₂CH₂COONa·6H₂O
MW: 270.14 g/mol
Melting Pt: 300
Density: 1.529 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149117
CAS Number: 6106-21-4
EINECS: 205-778-7
Merck Index: 13,08754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e2bd23508be53bfca204674aea2fd2cfea16837&vid=0e2bd23508be53bfca204674aea2fd2cfea16837&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR