Bis(2-ethylhexyl)sebacate ≥98.0%

Supplier: TCI America

S0025-500ML S0025-25ML
TCS0025-500MLEA 57.84 CAD
TCS0025-500ML TCS0025-025ML
Bis(2-ethylhexyl)sebacate ≥98.0%
Bis(2-ethylhexyl) sebacate

CAS Number: 122-62-3
MDL Number: MFCD00009497
Molecular Formula: C26H50O4
Molecular Weight: 426.68
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 377
Melting point (°C): -67
Flash Point (°C): 222
Specific Gravity (20/20): 0.91


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Di(2-ethylhexyl) Sebacate, Dioctyl Sebacate, Sebacic Acid Bis(2-ethylhexyl) Ester, Sebacic Acid Dioctyl Ester

Formula: C₂₆H₅₀O₄
Boiling Pt: 377 °C
Melting Pt: –67 °C
Density: 0.91 (20 °C)
Flash Pt: 222 °C
Storage Temperature: Ambient
MDL Number: MFCD00009497
CAS Number: 122-62-3
EINECS: 204-558-8
Merck Index: 13,01251
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fb2cf119e706869f7cfd145bf12a7650b77d074&vid=1fb2cf119e706869f7cfd145bf12a7650b77d074&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR