Butyl salicylate ≥99.0%

Supplier: TCI America

S0010-25ML S0010-500ML
TCS0010-025MLEA 44.24 CAD
TCS0010-025ML TCS0010-500ML
Butyl salicylate ≥99.0%
Butyl salicylate

CAS Number: 2052-14-4
MDL Number: MFCD00020038
Molecular Formula: C11H14O3
Molecular Weight: 194.23
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 260
Flash Point (°C): 134
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: Salicylic Acid Butyl Ester, 2-Hydroxybenzoic Acid Butyl Ester, Butyl 2-Hydroxybenzoate

Formula: C₁₁H₁₄O₃
Boiling Pt: 260 °C
Density: 1.07 (20 °C)
Flash Pt: 134 °C
MDL Number: MFCD00020038
CAS Number: 2052-14-4
EINECS: 218-142-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0a0c5159a7daf9c93044173a8e8d9c7aa6c427a&vid=c0a0c5159a7daf9c93044173a8e8d9c7aa6c427a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR