Sulfuric acid 0.255 N in aqueous solution (12.5 g/L) suitable for crude fiber analysis of food and feed

Supplier: Ricca Chemical

R8272000-4A R8272000-1A R8272000-20F
RC82721EA 167.48 CAD
RC82721 RC827232 RC82725
Sulfuric acid 0.255 N in aqueous solution (12.5 g/L) suitable for crude fiber analysis of food and feed
Sulfuric acid

AOAC for crude fiber.

Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 0.2550 ± 0.0005 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR