Volumetric Flasks with White Neck and [ST] Polyethylene Stopper, Class A, Hirschmann
Supplier: Hirschmann
282P25W 282P50W 282P100W
89085-146PK 164.37 CAD
89085-146 89085-148 89085-150
Volumetric Flasks with White Neck and [ST] Polyethylene Stopper, Class A, Hirschmann
Flasks Volumetric Flasks
Flasks include a [ST] polyethylene stopper, a fine white graduation line, and a marking spot
Meet ASTM E288, Class A requirements.
Flasks are designed with wide necks for easy pipet access. Graduated to contain. All flasks feature lot number and production year printed onto the vessel for QA purposes.