5-Sulfosalicylic acid 20% (w/v) in aqueous solution

Supplier: Ricca Chemical
Danger

Synonyms: 2-Hydroxy-5-sulphobenzoic acid, 2-Hydroxy-5-sulfobenzoic acid

R8130000-500C R8130000-1C
RC813016EA 235.42 CAD
RC813016 RC813032
5-Sulfosalicylic acid 20% (w/v) in aqueous solution
5-Sulfosalicylic acid solutions
Formula: HO₃SC₆H₃-2-(OH)CO₂H·2H₂O
Boiling Pt: ~ 100 °C
Density: 1.1000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00007508
CAS Number: 5965-83-3

Order Now

Specification Test Results

Appearance Clear, tannish-gold to light pink, odorless
Assay at 20 °C (traceable to NIST SRM 84) 20.00 ± 0.02 % (w/v)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR