PYREX® VISTA™ Volumetric Flasks with [ST] Stopper, Class A, Corning
Supplier: Corning
70640-10 70640-1L 70640-100 70640-2L 70640-200 70640-250 70640-25 70640-500 70640-50
89090-116CS 664.26 CAD
89090-116 89090-298 89090-304 89090-330 89090-334 89090-342 89090-354 89090-380 89090-392
PYREX® VISTA™ Volumetric Flasks with [ST] Stopper, Class A, Corning
Flasks Volumetric Flasks
PYREX® borosilicate glass, clear, with glass stoppers.
  • Flasks provide precise volume measurement
  • Flask neck tooled for No. 13 standard taper glass stoppers
  • Graduation lines are sharp and permanent
Vessels feature sealed necks tooled for standard taper stoppers and flat bottoms.
Certifications: ASTM E-694, E-544, E-288