5-Sulfosalicylic acid 3% (w/v) in aqueous solution

Supplier: Ricca Chemical
Warning

R8115000-500C R8115000-1C R8115000-4C
RC811516EA 97.96 CAD
RC811516 RC811532 RC8115-1
5-Sulfosalicylic acid 3% (w/v) in aqueous solution
5-Sulfosalicylic acid solutions
Formula: HO₃SC₆H₃-2-(OH)CO₂H·2H₂O
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0100 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00007508
CAS Number: 5965-83-3

Order Now

Specification Test Results

Appearance Clear, slight tannish gold to slight pink, odorless
Assay at 20 °C (traceable to NIST SRM 84) 3.00 ± 0.01 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR