Starch 1% (w/v) in aqueous solution for iodometric titrations mercury-free

Supplier: Ricca Chemical

R8050000-4A R8050000-10F R8050000-1A R8050000-20F R8050000-500A
RC80501EA 208.56 CAD
RC80501 RC805025 RC805032 RC80505 RC805016
Starch 1% (w/v) in aqueous solution for iodometric titrations mercury-free
Starch

Indicator. Translucent liquid. For general iodometric titrations.


  • Preserved with 0.01–0.2% salicylic acid
  • Mercury free.

Formula: (C₆H₁₀O₅)n
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-25-8
EINECS: 232-679-6

Order Now

Specification Test Results

Appearance Translucent, odorless
Suitability for Use Colorless (Iodine absent) - Blue (Iodine present)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR