Starch 0.5% (w/v) in aqueous solution for iodometric titrations mercury-free

Supplier: Ricca Chemical

R8000000-4A R8000000-500A R8000000-1A R8000000-20F
RC80001EA 221.2 CAD
RC80001 RC800016 RC800032 RC8000-5
Starch 0.5% (w/v) in aqueous solution for iodometric titrations mercury-free
Starch

Indicator. APHA. (Salicyclic Acid Preservative).

Formula: (C₆H₁₀O₅)n
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-25-8
EINECS: 232-679-6

Order Now

Specification Test Results

Appearance Translucent, odorless
Suitability for Use Colorless (Iodine absent) - Blue (Iodine present)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR