Sodium thiosulfate 0.0394 N aqueous solution Iodine Adsorption Number of Carbon Black

Supplier: Ricca Chemical

Synonyms: di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite

R7930000-4A R7930000-20F
RC79301EA 262.28 CAD
RC79301 RC79305
Sodium thiosulfate 0.0394 N aqueous solution Iodine Adsorption Number of Carbon Black
Sodium thiosulphate

ASTM D-1510, for iodine absorption number of carbon black.

Formula: Na₂S₂O₃
MW: 158.11 g/mol
Boiling Pt: 100
Melting Pt: 48
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5
Merck Index: 12,08844

Order Now

Specification Test Results

Appearance Clear, colorless, slight organic odor
Assay at 20 °C (traceable to NIST SRM 136) 0.03940 ± 0.00001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR