Sodium hydroxide : sodium thiosulfate (40:2.5) % (w/v) in aqueous solution for Kjeldahl nitrogen analysis using mercury catalyst
Supplier: Ricca Chemical
|
Danger
|
R7490000-4A
R7490000-10F
R7490000-20F
RC74901EA
206.98
CAD
RC74901
RC749025
RC74905
Sodium hydroxide : sodium thiosulfate (40:2.5) % (w/v) in aqueous solution for Kjeldahl nitrogen analysis using mercury catalyst
Sodium hydroxide : sodium thiosulfate
APHA and ASTM for organic nitrogen.
Boiling Pt:
~ 100 °C Density: 1.4000 g/cm³ Storage Temperature: Ambient |
UN:
1824 ADR: 8,II |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 136) | 2.5 ± 0.1 % (w/v) |
Assay at 20 °C (traceable to NIST SRM 723) | 40.0 ± 0.4 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...