Sodium hydroxide 1.00 Normal, Tightrant™ analytical reagent Tested in an ISO 17025 Accredited Facility
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Caustic soda
R7450000-4F
R7450000-10F
R7450000-1A
R7450000-20F
R7450000-4A
R7450000-4F
R7450000-120A
R7450000-20E1
RC7450-1CTEA
219.62
CAD
RC7450-1CT
RC745025
RC745032
RC74505
RC74501
CARC7450-1CT
CARC7450-4
RCR745000020E1
Sodium hydroxide 1.00 Normal, Tightrant™ analytical reagent Tested in an ISO 17025 Accredited Facility
Sodium hydroxide
Standardized at 25°C against chemicals whose certification is traceable to NIST.
Formula:
NaOH Melting Pt: ∼ 0°C Density: 1.04 Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,III |
Specification Test Results
Appearance | Colorless liquid |
Assay (vs. Potassium Hydrogen Phthalate/Phenolphthalein) | 0.999-1.001 N at 20°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...