KIMBLE® RAY-SORB® Amber Heavy Duty Wide Mouth Volumetric Flask, No Stopper, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
92822N-10 92822N-20 92822N-25 92822N-100 92822N-200 92822N-250 92822N-500 92822N-1000 92822N-2000 92822N-5 92822N-50
89079-308CS 631.92 CAD
89079-308 89079-310 89079-312 89079-316 89079-318 89079-320 89079-322 89079-324 89079-326 89079-306 89079-314
KIMBLE® RAY-SORB® Amber Heavy Duty Wide Mouth Volumetric Flask, No Stopper, DWK Life Sciences
Flasks Volumetric Flasks
Flasks are made of red-tinted glass that provides protection from UV light
All flasks are graduated to Class A volumetric tolerances. Flasks with 5–250mL capacity are calibrated to meet ASTM Specification E288.
Feature wide mouth for easy pipet access and heavy, uniform walls for strength, durability, and safety. Interior bottoms are flat, allowing for efficient stirring with standard stir bars. Large, permanent markings allow for quick, accurate identification. Sold without stoppers.