Sodium hydroxide 0.200 N (N/5)

Supplier: Ricca Chemical
Warning

R7360000-1A R7360000-20F R7360000-4A R7360000-500A
RC736032EA 90.06 CAD
RC736032 RC73605 RC73601 RCR7360000500A
Sodium hydroxide 0.200 N (N/5)
Sodium hydroxide
Formula: NaOH
Boiling Pt: ∼105 °C (1013 hPa)
Melting Pt: 0.0°C
Density: 1.01
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Potassium Hydrogen Phthalate/Phenolphthalein) 0.1999-0.2001 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR