Sodium hydroxide 0.100 N (N/10) in aqueous solution
Supplier: Ricca Chemical
R7350000-500A
R7350000-10F
R7350000-20F
R7350000-4A
R7350000-1A
RC735016EA
77.42
CAD
RC735016
RC735025
RC73505
RC73501
RC735032
Sodium hydroxide 0.100 N (N/10) in aqueous solution
Sodium hydroxide
Standardized at 25°C against chemicals whose certification is traceable to NIST.
Formula:
NaOH Density: 1 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,III |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 84) | 0.1000 ± 0.0001 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...