Sodium hydroxide 0.0500 N (N/20) analytical reagent

Supplier: Ricca Chemical

Synonyms: Caustic soda

R7330000-4A R7330000-1A R7330000-20F R7330000-500A
RC73301EA 189.6 CAD
RC73301 RC733032 RC73305 RCR7330000500A
Sodium hydroxide 0.0500 N (N/20) analytical reagent
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 1
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 III
Merck Index: 14,08627

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Potassium Hydrogen Phthalate/Phenolphthalein) 0.04995-0.05005 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR