Sodium hydroxide 0.0200 Normal (N/50) (1 ml = 1 mg CaCO₃), Tightrant™ analytical reagent Tested in an ISO 17025 Accredited Facility

Supplier: Ricca Chemical

Synonyms: Caustic soda

R7300000-4A R7300000-1A R7300000-20F R7300000-500A
RC73001EA 197.5 CAD
RC73001 RC730032 RCR7300000-20F RCR7300000500A
Sodium hydroxide 0.0200 Normal (N/50) (1 ml = 1 mg CaCO₃), Tightrant™ analytical reagent Tested in an ISO 17025 Accredited Facility
Sodium hydroxide

APHA for acidity.

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 1
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 III
Merck Index: 14,08627

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Potassium Hydrogen Phthalate/Phenolphthalein) 0.01995-0.02005 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR