Sodium hydroxide 10% (w/v) in aqueous solution (100 g/L)

Supplier: Ricca Chemical
Danger

R7260000-500A R7260000-1A R7260000-120A R7260000-4A R7260000-20F
RC726016EA 75.84 CAD
RC726016 RC726032 RC72604 RC7260-1 RCR7260000-20F
Sodium hydroxide 10% (w/v) in aqueous solution (100 g/L)
Sodium hydroxide
Formula: NaOH
Boiling Pt: ~100 °C
Melting Pt: ~0 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 9.96 - 10.04 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR