KIMAX® Volumetric Flasks with [ST] Polyethylene Stopper, Wide Mouth, Class A, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
92812P-20 92812P-25 92812P-100 92812P-200 92812P-500 92812P-1000 92812P-2000 92812P-5 92812P-10 92812P-50 92812P-250
89079-200CS 614.75 CAD
89079-200 89079-202 89079-206 89079-208 89079-212 89079-214 89079-216 89079-196 89079-198 89079-204 89079-210
KIMAX® Volumetric Flasks with [ST] Polyethylene Stopper, Wide Mouth, Class A, Kimble Chase
Flasks Volumetric Flasks
Flasks feature wide mouth for easy pipet access and heavy, uniform walls for strength, durability, and safety
All flasks are graduated to Class A volumetric tolerances. Flasks with 5–250mL capacity are calibrated to meet ASTM Specification E288.
Interior bottoms are flat, allowing for efficient stirring with standard stir bars. Large, permanent markings allow for quick, accurate identification.