Sodium chloride 5% (w/v) in aqueous solution

Supplier: Ricca Chemical

Synonyms: Saline, NaCl, Sea salt, Halite

R7215000-500A R7215000-1A R7215000-55E
RC721516EA 75.84 CAD
RC721516 RC721532 RC7215000-55E
Sodium chloride 5% (w/v) in aqueous solution
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Melting Pt: ∼ 0°C
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 5.000 ± 0.006 % (w/v)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR