Sodium chloride standard solution 1,00% (w/w) in aqueous solution analytical reagent
Supplier: Ricca Chemical
Synonyms:
Saline, NaCl, Sea salt, Halite
R7213000-4A
R7213000-500A
RC7213-1EA
407.64
CAD
RC7213-1
RC721316
Sodium chloride standard solution 1,00% (w/w) in aqueous solution analytical reagent
Sodium chloride
Formula:
ClNa Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 EINECS: 231-598-3 |
Specification Test Results
% (w/w) (traceable to NIST SRM 999) | 1.00 ± 0.01 % (w/w) |
Appearance | Clear, colorless, odorless |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...