KIMBLE® RAY-SORB® Amber Heavy Duty Wide Mouth Volumetric Flask, Glass Stopper, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
92822G-20 92822G-25 92822G-100 92822G-200 92822G-500 92822G-1000 92822G-2000 92822G-5 92822G-10 92822G-50
89079-244CS 682.07 CAD
89079-244 89079-246 89079-250 89079-252 89079-256 89079-258 89079-260 89079-240 89079-242 89079-248
KIMBLE® RAY-SORB® Amber Heavy Duty Wide Mouth Volumetric Flask, Glass Stopper, DWK Life Sciences
Flasks Volumetric Flasks
Flasks are made of red-tinted glass that provides protection from UV light
All flasks are graduated to Class A volumetric tolerances. Flasks with 5–250mL capacity are calibrated to meet ASTM Specification E288.
Feature wide mouth for easy pipet access and heavy, uniform walls for strength, durability, and safety. Interior bottoms are flat, allowing for efficient stirring with standard stir bars. Large, permanent markings allow for quick, accurate identification.