VWR® Glove Liners
Supplier: VWR International
94000-608 94000-610 94000-612 94000-614 94000-616 94000-618 94000-620 94000-622 10124-750 77392-160
94000-608CS 945.72 CAD
94000-608 94000-610 94000-612 94000-614 94000-616 94000-618 94000-620 94000-622 10124-750 77392-160
VWR® Glove Liners
Gloves
Ergonomically designed glove liners wick away moisture for comfortable use with disposable gloves.
  • Ambidextrous glove liners
  • Reduces perspiration and wicks away moisture
  • Low-lint, 100% 15 denier nylon composition
  • Sturdy, launderable, and reusable
  • Color-coded cuffs indicate size
  • Pre-laundered; will not shrink
Liners provide protection and comfort when used with latex, nitrile, and PVC gloves. Fully launderable for reusability and economy.
Packaging: Glove Liners are packaged 20 pair per bag, 15 bags per case. They are packed in clean polyethyleneMore Product Information