KIMAX® Volumetric Flasks with Color-Coded [ST] PTFE Stopper, Wide Mouth, Class A, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
92812F-5 92812F-20 92812F-25 92812F-200 92812F-250 92812F-500 92812F-2000 92812F-10 92812F-50 92812F-100 92812F-1000
89079-174CS 540.05 CAD
89079-174 89079-178 89079-180 89079-186 89079-188 89079-190 89079-194 89079-176 89079-182 89079-184 89079-192
KIMAX® Volumetric Flasks with Color-Coded [ST] PTFE Stopper, Wide Mouth, Class A, Kimble Chase
Flasks Volumetric Flasks
Flasks feature wide mouth for easy pipet access and heavy, uniform walls for strength, durability, and safety
All flasks are graduated to Class A volumetric tolerances. Flasks with 5–250mL capacity are calibrated to meet ASTM Specification E288.
Interior bottoms are flat, allowing for efficient stirring with standard stir bars. Large, permanent markings allow for quick, accurate identification.