Sodium chloride 0.85% (w/v) in aqueous solution

Supplier: Ricca Chemical

R7200000-4A R7200000-500A R7200000-10F R7200000-1A R7200000-20F
RC72001EA 121.66 CAD
RC72001 RC720016 RC720025 RC720032 RC72005
Sodium chloride 0.85% (w/v) in aqueous solution
Sodium chloride

Normal saline, sub-micron filtered, low particle count, nonsterile. Suitable for use as Blood Bank Saline.

Formula: NaCl
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 0.851 ± 0.001 % (w/v)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR