Sodium carbonate 1.00 N
Supplier: Ricca Chemical
|
Danger
|
R7190000-4A
R7190000-1A
R7190000-500A
RC71901EA
249.64
CAD
RC71901
RC719032
RCR7190000500A
Sodium carbonate 1.00 N
Sodium carbonate
Clear, colorless liquid. Less than 1% sodium carbonate. Irritant.
Formula:
Na₂CO₃ Melting Pt: ∼ 0°C Density: 1.1 Storage Temperature: Ambient |
MDL Number:
MFCD00003494 CAS Number: 497-19-8 EINECS: 207-838-8 |
|
Specification Test Results
Appearance | Colorless liquid |
Assay (vs. Sulfuric Acid/Methyl Orange) | 0.999-1.001 N at 20°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...