Silver nitrate 0.171 N in aqueous solution

Supplier: Ricca Chemical
Warning

R7003000-4B R7003000-1B R7003000-20F
RC70031EA 1880.2 CAD
RC70031 RC700332 RC70035
Silver nitrate 0.171 N in aqueous solution
Silver nitrate

For chloride. 1mL = 10mg NaCl.

Formula: AgNO₃
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 0.1710 ± 0.0001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR