KIMAX® Volumetric Flasks with [ST] Glass Stopper, Wide Mouth, Class A, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
92812G-5 92812G-20 92812G-25 92812G-100 92812G-200 92812G-250 92812G-500 92812G-2000 92812G-10 92812G-50 92812G-1000
89079-152CS 499.92 CAD
89079-152 89079-156 89079-158 89079-162 89079-164 89079-166 89079-168 89079-172 89079-154 89079-160 89079-170
KIMAX® Volumetric Flasks with [ST] Glass Stopper, Wide Mouth, Class A, Kimble Chase
Flasks Volumetric Flasks
Flasks feature wide mouth for easy pipet access and heavy, uniform walls for strength, durability, and safety
All flasks are graduated to Class A volumetric tolerances. Flasks with 5–250mL capacity are calibrated to meet ASTM Specification E288.
Interior bottoms are flat, allowing for efficient stirring with standard stir bars. Large, permanent markings allow for quick, accurate identification.