Perchloric acid 0.1 N in glacial acetic acid, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

BAKR5624-02 5624-02
CAJT5624-2CS 3491.45 CAD
JT5624-2 CAJT5624-2
Perchloric acid 0.1 N in glacial acetic acid, BAKER ANALYZED® volumetric solution, J.T.Baker®
Perchloric acid
Formula: HClO₄
MW: 100.46 g/mol
MDL Number: MFCD00011325
CAS Number: 7601-90-3
UN: 2920
ADR: 8,II
https://ca.vwr.com/stibo/web/std.lang.all/06/22/16640622.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality, For Information Only
Standardization at 25°C traceable to NIST Standard Reference Material.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR