Potassium hydroxide 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Potash caustic, Caustic potash

Management of Change (MOC) category = N

BAKR5603-03 5603-02
CAJT5603-3CS 2620.54 CAD
CAJT5603-3 JT5603-2
Potassium hydroxide 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Potassium hydroxide

Standardized at 25°C against chemicals whose certification is traceable to NIST. Lot standardization value, accurate to ±1 part per 1000, provided on label.

Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8 II
Merck Index: 12,07806

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.0995 - 0.1005
Chloride (Cl) ≤10 ppm
Nitrogen Compounds (as N) ≤1 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Trace Impurities - Iron (Fe) ≤0.5 ppm
Appearance Passes Test
Standardization at 25°C traceable to NIST Standard Reference Material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR