Volumetric Flasks with [ST] Glass Stopper, Class A, Amber, Hirschmann
Supplier: Hirschmann
264-G-10 264-G-25 264-G-50 264-G-100 264-G-200 264-G-250 264-G-500 264-G-1000
89025-782PK 247.85 CAD
89025-782 89025-784 89025-786 89025-788 89025-790 89025-792 89025-794 89025-796
Volumetric Flasks with [ST] Glass Stopper, Class A, Amber, Hirschmann
Flasks Volumetric Flasks
Flasks feature amber-colored glass for the protection of light-sensitive contents, a fine white graduation line, and marking spot
Meet ASTM E288, Class A requirements.
Graduated to contain. Flasks include a [ST] glass stopper.