Sodium hydroxide 10.0 N aqueous solution, StandARd® Volumetric, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Caustic soda

M.O.C. Category NI

MACRH385-05 MACRH385-07
CAMKH38505EA 302.82 CAD
CAMKH38505 CAMKH38507
Sodium hydroxide 10.0 N aqueous solution, StandARd® Volumetric, Macron Fine Chemicals™
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Protect from air to avoid absorption of carbon dioxide.
Normality 9.95 - 10.05
Identification Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR