Spectra® Fiber Cut-Resistant Glove Liners, Wells Lamont
Supplier: Wells Lamont
Spectra®
M214L M214M M214S M214XS M214XL
32935-487BG 642 CAD
32935-487 32935-485 32935-483 32935-509 32935-481
Spectra® Fiber Cut-Resistant Glove Liners, Wells Lamont
Gloves
An ultrahigh molecular weight polyethylene fiber provides moderate cut resistance with high reusability.
  • Gloves can be used outside
Gloves are lightweight and have a low coefficient of friction, which allows sharp objects to slide across the surface.